Bromate-Bromide solution 0.250 M (0.500 N)
Supplier: Ricca Chemical
|
Danger
|
R1172000-4C
R1172000-1C
RC11721EA
505.6
CAD
RC11721
RC117232
Bromate-Bromide solution 0.250 M (0.500 N)
Bromate-Bromide
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 136) | 0.5000 ± 0.0002 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...