Bromate-Bromide solution 0.0500 N (N/20)
Supplier: Ricca Chemical
|
Danger
|
R1168000-1C
R1168000-4C
RC1168-32EA
235.42
CAD
RC1168-32
RC1168-1
Bromate-Bromide solution 0.0500 N (N/20)
Bromate-Bromide
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 136) | 0.05000 ± 0.00002 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...