Borax 1% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger

R1050000-500A R1050000-1A
RC105016EA 96.38 CAD
RC105016 RC1050-32
Borax 1% (w/v) in aqueous solution
di-Sodium tetraborate decahydrate
Formula: Na₂B₄O₇·10H₂O
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0085 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149193
CAS Number: 1303-96-4

Order Now

Specification Test Results

Appearance Clear, colorless, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR