Sealed Sample Tubes, Omni International
Supplier: Omni International
00-2015-25 00-2050-25
CA11010-556PK 476.8 CAD
CA11010-556 CA11010-558 CA11010-562
Sealed Sample Tubes, Omni International
Tubes Test Tubes
Sealed sample tubes are used with clear Omni Tip™ plastic generator probes to reduce harmful airborne contaminants.
  • Sealed tubes are designed as a secondary means of protection
Always use appropriate measures when processing pathogens.
Ordering information: Omni Tip™ Plastic Generator Probes are sold separately.
Order Now
SPECIFICATIONS
Environmentally Preferable