Weatherproof Flammable Safety Cabinets, SECURALL®
Supplier: Securall Cabinets
A130WP1 A330WP1 A145WP1 A345WP1 A160WP1 A360WP1 A190WP1 A390WP1
89237-744EA 4547.81 CAD
89237-744 89237-746 89237-748 89237-750 89237-752 89237-754 89237-756 89237-758
Weatherproof Flammable Safety Cabinets, SECURALL®
Furniture Storage Cabinets
Double-wall weatherproof cabinets are all-welded, 18-gauge construction using galvannealed steel and painted with a weather-resistant, impact-resistant urethane finish that is also resistant to ultra-violet rays.
All Cabinets STANDARD Features a sloped roof with a 2³/₄" overhang along with interior and exterior astragals to prevent rain penetration. Weatherproof cabinets come standard with corrosion resistant flush-mounted stainless steel locking handle, but may be upgraded to galvanized high security hasp lock. Easily move these cabinets to the appropriate work location using built-inMore Product Information