Bio Hazard Shields for Laboratory work, S-Curve Technologies
Supplier: S-Curve Technologies
BS-1010 BS-2016 BS-1212
14007-142CS 585.48 CAD
14007-142 14007-140 14007-138
Bio Hazard Shields for Laboratory work, S-Curve Technologies
Shields Safety Shields
Bio-hazard shields offer worker protection in the lab
3/8" clear acrylic with non-slip pads on the bottom.