Sterile IPA 70% Spray, ITW Texwipe
Supplier: ITW Texwipe
TX3270 TX3273
CATWTX3270CS 1635.81 CAD
CATWTX3270 CATWTX3273
Sterile IPA 70% Spray, ITW Texwipe
Cleaning Agents Disinfectants
This Sterile 70% Isopropanol (IPA) spray bottle ensures added sterility by eliminating the handling and assembly usually associated with IPA Trigger sprays
Choose a stream delivery for precise application or coarse spray for larger application areas. Fully assembled, ready-to-use applicator/container; no unpacking, handling or attachment of a separate spray head is required.
Meets FDA guideline for medical device bacterial endotoxin limit of not more than 20 EU/device.
IPA Quality meets requirements of USP Isopropyl Alcohol. Water Quality meets requirements of USP Purified Water.
Contains 70% by volume USP-grade Isopropanol and 30% USP-purified water. To further ensure sterility, this solution is submicron filtered, filled into cleaned containers, double-bagged and gamma-irradiated. Each bottle is lot coded with an expiration date for easy recordkeeping.
Ordering information: Each bottle includes a ready-to-use trigger sprayer and a set of 5 color indicator dots for easy identification. Certificate of Analysis provided with every lot.