N,N-Dimethylacetamide ≥98% (by GC), Supelco®

Supplier: MilliporeSigma
Danger

DX1545-3 DX1545-30
CADX1545-3EA 782 CAD
CADX1545-3 CADX1545-30
N,N-Dimethylacetamide ≥98% (by GC), Supelco®
N,N-Dimethylacetamide
Formula: CH₃CON(CH₃)₂
Boiling Pt: 165…166 °C (1013 hPa)
Melting Pt: –20 °C
Density: 0.94 g/cm3 (20 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
UN: 1993
ADR: 6.1,III
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 98% min
Color (APHA) 30 max
Form Clear liquid
Infrared spectrum Conforms to standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR