Sodium octanoate 99.0-101.0% NF, Ph. Eur., SAFC®

Supplier: MilliporeSigma
Warning

Synonyms: Octanoic acid sodium acid, Sodium caprylate, Caprylic acid sodium acid, Sodium n-Octanoate

8.17081.1000 8.17081.9010 8.17081.9025 8.17081.9010
CA8.17081.1000CS 4964.5 CAD
CA8.17081.1000 CA8.17081.9010 CA8.17081.9025 EM8.17081.9010
Sodium octanoate 99.0-101.0% NF, Ph. Eur., SAFC®
Sodium octanoate
Formula: C₈H₁₅NaO₂
MW: 166.2 g/mol
Melting Pt: 225…355 °C
Density: 0.9949 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00058511
CAS Number: 1984-06-1
EINECS: 217-850-5

Order Now

Specification Test Results

Assay (HClO4-titration, anhydrous) 99.0-101.0%
Appearance Almost white fine crystal powder
Appearance of solution (10%, water) Clear and colorless to almost colorless
Endotoxins 20U/g max
Heavy metals (as Pb) 0.0005% max
Identity Passes test
pH (10%, water) at 25°C 10.5 max
Related Substance (GC) largest single impurity 0.3% max
Related-Substance (GC) Sum of all impurity 0.5% max
Residual Solvents (Ph.Eur./ICH) Excluded by production process
Water 3.0% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR