Copper(II) sulfate pentahydrate, crystals, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, Extra Pure, SAFC®
Supplier: MilliporeSigma
|
Danger
|
1.02787.1000
1.02787.5000
1.02787.9050
CA1.02787.1000CS
3445.31
CAD
CA1.02787.1000
CA1.02787.5000
CA1.02787.9050
Copper(II) sulfate pentahydrate, crystals, EMPROVE® ESSENTIAL BP, Ph. Eur., USP, Extra Pure, SAFC®
Copper (II) sulphate pentahydrate
Formula:
CuSO₄·5H₂O Density: 2.284 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149681 CAS Number: 7758-99-8 EINECS: 231-847-6 UN: 3077 ADR: 9,III REACH: 01-2119520566-40 Merck Index: 13,02682 |
Specification Test Results
Assay (dried basis) | 98.5-100.5% |
Assay (iodometric) | 99.0-100.5% |
Appearance of solution | To pass test |
Calcium (Ca) | 0.005% max |
Chloride (Cl) | 0.01% max |
Identity | Passes Test |
Iron (Fe) | 0.003% max |
Lead (Pb) | 0.005% max |
Loss on drying (250°C) | 35.0-36.5% |
Nickel (Ni) | 0.005% max |
Organic volatile impurities (according to USP) | Conforms |
Potassium (K) | 0.01% max |
Residual Solvents (Ph.Eur./ICH) | Excluded by production process |
Sodium (Na) | 0.02% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...