Sorbic acid, EMPROVE® ESSENTIAL BP, FCC, NF, Ph. Eur., E200, SAFC®

Supplier: MilliporeSigma
Warning

Synonyms: (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid

1.00662.5000 1.00662.9025 1.00662.5000
CA1.00662.5000CS 1695.62 CAD
CA1.00662.5000 CA1.00662.9025 EM1.00662.5000
Sorbic acid, EMPROVE® ESSENTIAL BP, FCC, NF, Ph. Eur., E200, SAFC®
Sorbic acid
Formula: CH₃CH=CHCH=CHCOOH
MW: 112.13 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 132…135 °C
Density: 1.204 g/cm³ (20 °C)
Flash Pt: 127 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002703
CAS Number: 110-44-1
EINECS: 203-768-7
Merck Index: 13,08793
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=440c50b4fc52d9ff0b52afa7ba596c860f95e0e7&vid=f9b0ff409010208781696e9ca3314c02aaf1af36&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (acidimetric; calc. on anhydrous basis) 99.0-101.0%
Aldehydes (As Acetaldehyde) 0.15% max
Appearance of solution (5%; ethanol 96%) Passes
Arsenic (As) 0.0003% max
Cadmium (Cd) 0.001% max
Copper (Cu) 0.001% max
Heavy metals (as Pb) 0.0010% max
Identification Passes
Lead (Pb) 0.0002% max
Loss on drying 0.5% max
Melting range, beginning 133°C min
Melting range, end 135°C max
Mercury (Hg) 0.0001% max
Organic volatile impurities (according to NF) Passes
Other residual solvents (Ph.Eur/USP/ICH) Excluded by manufacturing process
Residual solvents (Ph.Eur/USP/ICH) of class 3 0.5% max
Sulfated ash 0.2% max
Thermal stability (105°C) No coloration
Water 0.5% max
Zinc (Zn) 0.001% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR