Omni Mixer High Power Homogenizer, Omni International, Inc.
Supplier: Omni International
17105
CA101099-998EA 18159.82 CAD
CA101099-998
Omni Mixer High Power Homogenizer, Omni International, Inc.
Homogenizers Handheld Homogenizers
The Omni Mixer Homogenizer is a versatile system that is well suited for laboratory & industrial processing applications such as Food, Cosmetics, Chemicals, Petrochemicals, Dyes and Waste Products.
Blade assemblies for mixing and rotor-stator generator probes for homogenization offer flexibility for blending, suspensions, dispersions, disintegrating, and emulsifications. Speed ranges of 500 to 20,000rpm allow for gentle mixing or higher speed homogenization type applications.
A variety of Generator Probes and Aerosol Sealed Chamber Assemblies are available to be used with the Omni Mixer Homogenizer. Sold seperately.
Certifications: CE approved.
Ordering information: Includes 600 watt variable speed motor, motor stand assembly, generator probe adapter, tool kit and instruction manual. Generator probes and chamber assemblies sold separately.