Omni Prep Multi-Sample Homogenizer, Omni International, Inc.
Supplier: Omni International
06-021
CA11010-682EA 100824.22 CAD
CA11010-682
Omni Prep Multi-Sample Homogenizer, Omni International, Inc.
Homogenizers Handheld Homogenizers
The innovative Omni Prep Multi-Sample Laboratory Homogenizer is specifically designed to streamline the labor and time intensive homogenization process and to make it more comfortable for the user
A specially designed oscillating sample rack holds up to 6 tubes from 2 mL to 50 mL in size. Microprocessor control allows the user to program time (to 10 minutes) and speed (500 - 30,000 rpm). Sample volumes of 250 µL to 30 mL can be homogenized using 5, 7 or 10 mm stainless steel generator probes or with Omni Tip™ Plastic Homogenizer Probes. Omni Tips completely eliminate cross-contamination as well as the time consuming cleaning step associated with use of stainless steel generator probes. They may be disposed of, reused, or autoclaved.
The Omni Prep is also suitable for sharing. As with a multi-sample centrifuge, an Omni Prep in the laboratory can elevate productivity and dramatically improve workflow. It is small enough to fit in most laminar flow cabinets. An optional ice tray is available to homogenize sensitive samples.
The Omni Prep features our new Whisper Drive Technology – a brushless motor that is virtually maintenance free, is ultra quiet during operation, and uses about 10% of the power required to process 6 samples individually. Unlike brush motors, since the motor holds its processing speed to +/- 1% accuracy throughout the homogenization cycle, Whisper Drive Technology guarantees accuracy and repeatability regardless of fluctuations in sample size or sample viscosity.
Ordering information: The Omni Prep is a customizable homogenizing system. At least 1 processing rack, 3 tube holders, and Omni Tip™ Plastic Probes are required to operate the Omni Prep.