VWR® Carboys with Versatile Caps, PP
Supplier: VWR International
89170-782 89170-784 89170-786 89170-790 89170-792 89170-794 89170-798 89170-800 89170-802 89170-806 89199-166 89217-554
89170-782EA 306.62 CAD
89170-782 89170-784 89170-786 89170-790 89170-792 89170-794 89170-798 89170-800 89170-802 89170-806 89199-166 89217-554
VWR® Carboys with Versatile Caps, PP
Bottles Carboys
VWR® polypropylene carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use.
  • Polypropylene construction
  • Translucent, autoclavable resin molding
The distinctive rectangular shape saves valuable bench space.
VWR® polypropylene carboys are molded from a translucent USP Class VI, FDA grade, autoclavable resin, which offers good chemical resistance and heat stability. Polypropylene carboys, as well as all versatile caps, adapters, versatile barbs, and spigots may be autoclaved. The recommended autoclaving cycle is 20 minutes at 15 psi. and 121 °C (250 °F).
Made in the USA.
Large handles on top of the container and molded handles in the base provide a full, secure grip to make lifting, carrying, and pouring easy. Specified models are available with a 120 mm wide neck, which allows the user to reach inside the carboy for easier cleaning. Leakproof polypropylene versatile caps offer the ultimate in connection flexibility. Open caps (sold separately) are compatible with interchangeable adapters, which feature a wide variety of top-connections, including molded-in hose barbs and quick-connect hose barbs. All carboys have their material and volume clearly imprinted on the outside of the bottle, as well as large, easy to read metric graduation marks which are certified to ±5% accuracy.