Ward's® Digital Slides: Advanced Placement Biology Set
Supplier: MASTERDATA QL HOLD 2
DEFAULT VPN DEFAULT VPN
470191-054EA 412.29 CAD
470191-054 470191-108
Ward's® Digital Slides: Advanced Placement Biology Set
Slides Prepared Slides
Ward's Digital Slides Powered By Motic®
  • Aligned To Accompany The AP Biology Big Ideas
  • Scan, Pan , Zoom In, And Focus Into Slides Just Like On A Microscope
  • Capture Images
  • Share Images With Students To Compare
Ward’s Digital Slides Powered by Motic® Hand-Prepared Quality Meets Digital Innovation Introducing the first and only digital slides collection of its kind, available at an affordable price for middle school, high school, and introductory college levels. Ward’s Digital Slides enhance traditional biology and microscopy labs through deeper analysis of specimens and structures, customized specimen interaction, and increased user control with every slide. Scanned by Revolutionary Motic® Technology Our vast collection of hand-prepared microscope slides has been digitally scanned by Motic®, the leaders in digital microscopy technology. This exclusive 3D scanning process captures specimen images at multiple magnifications and depths of field. The result is a crisp, high-resolution digital image that clearly depicts intricate details of structures at every level. Aligned with Common Science Standards Complete digital slides libraries are organized by curriculum topic, and aligned with common science standards and popular college textbooks. Complete Digital Slides Database at Your Fingertips Choose your Ward’s Digital Slides set and enjoy access to a full library of slides organized by curriculum topic. Measure structures on screen, navigate within thumbnails, delineate regions of interest, and capture images for further study. Exclusive Imaging Software Enables Customized Interaction Ward’s Digital Slides Image Interface by Motic® allows you to view, annotate, and save captured images from your slides library. Use to customize your lessons and presentations to suit your teaching needs, deepen student understanding, enhance student assessments, and much more.
System requirements:
Windows XP or higher operating system, Intel-based processor with 1GHz processor, 2GB RAM, 250MB hard disk space.
Ordering information: With Ward’s Digital Slides you can: Zoom up to 1000xMeasure structures on screen Navigate within layers of specimen along X, Y, and fine-focus Z-stacked axis Adjust fine focus of thick specimens along the Z-stacked axis captured by exclusive Motic 3D scanning process Capture and save images for annotation and further study. Enhance AP Biology investigations and deepen student understanding with this set of 107 vivid slide images selected to correspond to the Four Big Ideas and enduring understandings as outlined in the College Board AP Biology curriculum framework. Slides are organized within the slide library by curriculum standard and topic to allow quick navigation to the slide and concept you wish to teach. Purchase is for 12 months of cloud access and can be renewed.