Ward's® Digital Slides: Introductory Biology Digital Slide Set
Supplier: Avantor
470189-718EA 347.79 CAD
470189-718 470189-720
Ward's® Digital Slides: Introductory Biology Digital Slide Set
Slides Prepared Slides
With Ward's Digital Slides Powered By Motic® you can:
  • ScanandZoomupto1000X
  • Measure structures on screen
  • Navigate within layers of specimen along X,Y, and fine-focus Z axis
  • Adjust fine focus through thick specimens using the Z axis
  • Annotate slides on-screen for presentations and lectures
  • Capture and save images for student homework or pre-work
Our vast collection of microscope slides with over50 years of custom slide preparation experience is now available digitally. Enhance hands-on biology and microscopy labs through deeper analysis of specimens and structures, customized interaction, and increased user control.
Budding yeast/Fungi Saccharomyces (WM) 912501
Giardia lamblia-Trophozoites / Archaezoa (WM) 924233
Mixed protist/protista 920005
Ectocarpus / chromista (WM) Slide 911202
Amphioxus (WM) Slide chordate 923013
Arabidopsis wm 918125
ls zebrafish female 923133
Volvox-Sexual Stage (WM) Slide 910501
Paramecium-Fission (WM) Slide 920420
Spirogyra scalariform conjugation 910466
Saprolegnia-Sexual Stages (WM) 912122
Chlamydomonas-Flagella (WM) FS&FG 910270
Chara (Stonewort) (WM) Slide 910252
Psilotum Stem-Sporangia (LS) QS 914520
Equisetum Mature Strobilus (CS) QS 914818
Marchantia Thallus & Cupule (CS) 914042
Marchantia Antheridia (LS)QS Slide 914043
Marchantia Young Sporophyte (MLS) 914047
Marchantia Archegonia (LS)QS Slide 914045
Fern Prothallium-Monoecious (WM) 914862
Pinus strobus 5-needle Type (CS) 916503
Pine Ovule-Mature Archegonium (LS) 916544
Mixed Pollen (20 types) 917002
Tobacco Flower (LS) QS Slide 918056
Penicillium sp. 912471
Mushroom Anatomy-Coprinus (CS) QS 913211
Leucosolenia (Sponge) (WM) Slide 920568
Hydra Adult With Bud (WM) Slide 920651
Obelia Hydroids (WM) Slide 920730
Jellyfish Medusa 920779
Planaria Plain (WM) Slide 920820
Sclerids in Pear 931152
Idioblasts 931158
Trichomes 931210
Starch Grains /parenchyma (SECT) QS slide 931212
Wood Fibers (WM) WS SLide 931214
Casparian Strip (CS) QS Slide 931218
Collenchyma (CS) QS Slide 931220
Sclerenchyma in a Stem (CS) QS 931226
Sieve Plates (CS) QS Slide 931228
Tracheids in Herbaceous Stem, (LS) 931230
Lilium Leaf Epidermis (WM) FS&FG 917206
Coleus Stem Tip (LS) QS Slide 917833
Dianthus-Entire Leaf (CS) QS Slide 917882
Plasmodesmata (SECT) Slide 918090
Tilia 2-Year Old Stem (CS)QS Slide 918307
Zea Stem (LS) QS Slide 917450
Helianthus Older Stem (CS) QS 917914
Allium Mitosis (LS) IH&OG Slide 917040
Ranunculus root/dicot (CS) QS 918142
Ranunculus Root Mature(CS) QS Slide 918140
Ranunculus flower bud/dicot 918147
Ligustrum Leaf (CS) QS Slide 917940
Capsella embryo/ dicot (LS) QS 917808
Pigmented Epithelium(WM) Slide 923644
Frog Blood /connective tissue (SM) GS Slide 923640
Chondroid Tissue /connective (SECT) H&E Slide 933219
Frog Heart 923668
Frog Artery, Vein, Nerve (epithelium tissue in circulatory system, nerve tissue in nervous system) 923671
Mouse Tail (CS) H&E Slide 933321
Hydra-General Structure (CS) H&E 920632
Hydra Plain (WM) Slide 920630
Earthworm Intestinal Region (CS) 921800
Bird intestine 923811
Bird crop gizzard 923812
Mamal digestive system composite 934523
Cow rumen 934501
Cow reticulum 934502
Cow Omasum 934503
Cow abomasum 934504
Ectotrophic Mycorrhiza (CS) QS 919810
Lichen-Mycobiont (WM) Slide 913950
Termite Flagellates (WM) Slide 924260
Female anopheles mosquito /malaria 926521
Plasmodium malariae in human blood 924630
Plasmodium in liver 924701
Just like Google Earth™ for your microscope slides!
Ordering information: System requires Windows XP or higher operating system, Intel-based processor with 1GHz processor, 2GB RAM, 250MB hard disk space. Purchase is for 12 months of cloud access and can be renewed.