Dialysis Tubing Closure
Supplier: WELOC USA LLC
PA50
470206-374CS 12053.65 CAD
470206-374
Dialysis Tubing Closure
Educational Materials Biology Educational Materials Human Biology Learning Activities
Provides an Optimum Seal Every Time.
  • For use with tubing up to 50 mm in flat width
  • Secure, long-lasting, reusable
  • Eliminates the need to tie the dialysis tubing
The locking mechanism helps to seal the tubing and minimizes the risk of leakage. Made from DuPont Nylon PA6.6, it's the ultimate option when the optimum seal is required. 65 mm total length.