Innovating Science® Green Chemistry: The Production of Biodiesel
Supplier: ALDON CORP SE
Innovating Science®
IS9500 IS9500-SGL
470233-568KT 130.1 CAD
470231-564 470233-568
Innovating Science® Green Chemistry: The Production of Biodiesel
Educational Materials Chemistry Educational Materials Green Chemistry Learning Activities
Teach your students how biodiesel is made.
  • Green chemistry in a real-life experimental setting
  • Aligned to NGSS Practices
  • Performance Expectations: MS-PS1-2, MS-PS1-3, HS-PS1-2
  • Disciplinary Core Ideas: PS1.A, PS1.B
  • Cross Cutting Concepts: Patterns, structure and function
  • Engineering Practices: Analyzing and interpreting data; obtaining, evaluating, and communicating information
In this activity, students will be performing a two-phase process to produce small batches of crude biodiesel. The crude biodiesel produced is of sufficient quality for use in the demonstration of the burning qualities of both biodiesel and vegetable oil. Included is an optional small-scale exercise where the students will use a washing procedure to experience the full process of producing biodiesel to meet quality levels necessary for use in vehicles. Kit contains enough materials for 15 groups of students.
Includes:
Vegetable oil
Methyl alcohol
Potassium hydroxide
Microburners
Teacher's guide and student copymasters
Delivery information: Instructor's Manual and SDS included.