Organic Chemistry Glassware Set
Supplier: United Scientific Supplies
ORGKIT-09 ORGKIT-16
470136-742KT 341.5 CAD
470136-742 470136-706
Organic Chemistry Glassware Set
Educational Materials Chemistry Educational Materials Microchemistry Learning Activities
This set features the must-haves for organic chemistry labs.
  • Set of 9 or 16 Items Available
  • All Glassware Is Jointed
  • Sturdy, Form-Fitting Case
  • Standard Version Covers Essential Organic Chemistry Requirements
  • Deluxe Set Works with Preparations up to 150 g
The standard version is designed to cover essential requirements for organic chemistry and is suitable for preparations up to 30 g. The deluxe set is suitable for preparations up to 150 g.
It's available in a set of nine or 16 items, and both sets come in a sturdy, form-fitting case.