Omni Tip™ Homogenizing Kits, Omni International, Inc.
Supplier: Omni International
TH115-PCR5D TH115-PCRH TH115-PCR10D TH115-PCR7H TH115-PCRD TH115-PCR10H TH115-PCR5H TH115-PCR TH115-PCR7D
CA11010-520EA 14406.41 CAD
CA11010-520 CA11010-530 CA11010-518 CA11010-526 CA11010-514 CA11010-528 CA11010-516 CA11010-522 CA11010-524
Omni Tip™ Homogenizing Kits, Omni International, Inc.
Homogenizers Handheld Homogenizers
Omni Tip™ Homogenizing Kits include everything you need to homogenize
Patented Omni Tip™ Plastic Homogenizer Probes are designed to completely avoid cross-contamination between samples during homogenization. Engineered from a durable plastic, they come in two different configurations for soft tissue and hard tissue homogenizing. Omni Tips are available in a standard size of 7mm (Diameter) x 110mm (Length). The outer tube of Omni Tips is made of Lexan 144R, a polycarbonate. The inner shafts are made of Ultem 1000, a polyetherimide.
They come complete with an Omni TH Tissue Homogenizer motor, Omni Tip™ adapter, and 12 Omni Tip™ Plastic Generator Probes. These kits can be used to process sample sizes from 250µL to 30mL with Omni Tips & <200µL to 100mL with optional stainless steel generator probes.
Ordering information: Includes an Omni TH Motor, Omni Tip™ Adapter, 12 Soft or 12 Hard Tissue Omni Tips™, stand clamp, convenient storage case and instruction manual.