Omni TH Handheld Tissue Homogenizer, Omni International, Inc.
Supplier: Omni International
TH220 TH115
CA11005-598EA 5586.93 CAD
CA11005-598 CA11011-614
Omni TH Handheld Tissue Homogenizer, Omni International, Inc.
Homogenizers Handheld Homogenizers
The Omni TH is a powerful Tissue Homogenizer designed to handle a wide range of laboratory samples from <200 µl to 100ml.
The Omni TH will accommodate a wide range of lab applications such as tissue homogenization, emulsions and suspensions, cell disruption, protein extraction and medical research. It is lightweight enough to be hand-held and can also be mounted to a motor stand for lengthier processing.
Note: Stainless Steel Generator Probes and Omni Tip™ Plastic Probes are sold separately.
The TH can usually process a tissue sample in less than 30 seconds and is compatible with a wide variety of generator probes for flexibility in application requirements.
Certifications: CE Approved.
Ordering information: Includes TH motor, 1-year motor warranty, tool kit, and instruction manual.