SP Bel-Art Precisionware® Bottles, Wide Mouth, HDPE, Bel-Art Products, a part of SP
Supplier: Bel-Art
F10625-0018 F10625-0005 F10625-0006 10625-9007 10625-9005 10625-9018 F10625-0007 10625-9006
12000-121CS 564.67 CAD
12000-121 16126-759 47748-112 47745-338 47748-114 16126-760 47750-620 47748-110
SP Bel-Art Precisionware® Bottles, Wide Mouth, HDPE, Bel-Art Products, a part of SP
Bottles Plastic Bottles
Rigid high-density polyethylene, laboratory/pharmaceutical grade bottles are leakproof and supplied with matched polypropylene closures.
  • Rigid HDPE construction
  • Polypropylene closures
  • Leakproof design
  • Recycling code molded into the base
Constructed from FDA compliant material, the wide mouth design allows for easy filling and pouring. The design also simplifies the packaging and storing of liquids and powders. Bottles 250 ml and larger feature a recycling code molded into the base.