SP Bel-Art Precisionware® Bottles, Narrow Mouth, HDPE, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
10620-9007 10620-9008 10620-9015 10620-9016 F10620-0007 F10620-0015 F10620-0016
47748-058CS 703.86 CAD
47748-058 47748-060 47748-062 47748-064 47750-604 47750-608 47750-610
SP Bel-Art Precisionware® Bottles, Narrow Mouth, HDPE, Bel-Art Products, a part of SP
Bottles Plastic Bottles
Rigid high-density polyethylene, laboratory/pharmaceutical grade bottles have matched polypropylene closures.
  • Rigid HDPE construction
  • Polypropylene closures
  • Leakproof design
  • Recycling code molded into the base
The leakproof design makes them ideal for packaging and storage of liquids and powders. Bottles 250 ml and larger feature a recycling code molded into the base.