Potassium hydroxide 10% w/v aqueous solution, StandARd®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Potash caustic, Caustic potash

M.O.C. Category N

H320-05 H320-01
MKH32005CS 406.46 CAD
MKH32005 MKH32001
Potassium hydroxide 10% w/v aqueous solution, StandARd®, Macron Fine Chemicals™
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (KOH)(g/100 mL) 9.5 - 10.5
Identification Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR