Ammonium sulfate ≥99%, GenAR® for biotechnology, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

Management of Change (MOC) category = N

MACR7725-65 MACR7725-88 MACR7725-22
CAMK772565EA 2612.96 CAD
CAMK772565 CAMK772588 CAMK7725-22
Ammonium sulfate ≥99%, GenAR® for biotechnology, Macron Fine Chemicals™
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
Merck Index: 13,00559

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay ≥99.0 %
Solubility Passes Test
ACS - Arsenic (As) ≤0.00002 %
ACS - Chloride (Cl) ≤0.0002 %
ACS - Heavy Metals (as Pb) ≤0.0005 %
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤0.0003 %
ACS - Nitrate (NO₃) ≤0.0002 %
ACS - Phosphate (PO₄) ≤0.0005 %
ACS - Residue on Ignition ≤0.005 %
ACS - pH of 5% Solution at 25°C 5.0 - 6.0
Copper (Cu) ≤1 ppm
Zinc (Zn) ≤1.000 ppm
Enzyme Activity - DNase Activity Passes Test
Enzyme Activity - RNase Activity Passes Test
Enzyme Activity - Protease Activity Passes Test
Endotoxin Concentration (EU/g) ≤2.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR