Calcium hydrogen orthophosphate, anhydrous 98.0-103.0%, powder USP, FCC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = R

MACR4264-20 MACR4264-24
CAMK426420EA 4590.14 CAD
CAMK426420 CAMK426424
Calcium hydrogen orthophosphate, anhydrous 98.0-103.0%, powder USP, FCC, Macron Fine Chemicals™
Calcium hydrogen orthophosphate
Formula: CaHPO₄
Melting Pt: 370 °C
Density: 2.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010909
CAS Number: 7757-93-9
EINECS: 231-826-1
Merck Index: 13,01697

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets F.C.C. Requirements
Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
USP - Assay (CaHPO₄) 97.5 - 102.5 %
USP - Loss on Ignition 6.6 - 8.7 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Acid-Insoluble Substances ≤0.2 %
USP - Carbonate (CO₃) Passes Test
USP - Chloride (Cl) ≤0.25 %
USP - Fluoride (F) ≤0.005 %
USP - Sulfate (SO₄) ≤0.5 %
USP - Arsenic (As) ≤3 ppm
USP - Barium (Ba) Passes Test
FCC - Assay (CaHPO₄) 98.0 - 103.0 %
FCC - Loss on Ignition 7.0 - 8.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Fluoride (F) ≤0.005 %
FCC - Arsenic (As) ≤3 ppm
FCC - Lead (Pb) ≤2 ppm
Appearance Passes Test
Solubility Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR