Hydrochloric acid 10% aqueous solution NF, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

2608-45 MACR2608-08 MACR2608-22 MACR2608-02 MACR2608-27
MK260845CS 1351.17 CAD
MK260845 CAMK2608-08 CAMK260822 CAMK260802 CAMK2608-27
Hydrochloric acid 10% aqueous solution NF, Macron Fine Chemicals™
Hydrochloric acid
Formula: HCl
MW: 36.46
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (HCl)(g/100ml) 9.5 - 10.5
NF - Identification (Cl) Passes Test
NF - Residue on Ignition ≤2 mg
NF - Free Bromine or Chlorine Passes Test
NF - Sulfate (SO₄) Passes Test
NF - Sulfite (SO₃) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR