Guanidinium hydrochloride 99,5, GenAR® Biotechnology, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

M.O.C. Category R

MACR0506-06 MACR0506-04
CAMK050606EA 3349.35 CAD
CAMK050606 CAMK050604
Guanidinium hydrochloride 99,5, GenAR® Biotechnology, Macron Fine Chemicals™
Guanidine hydrochloride
Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Appearance Passes Test
Water-Insoluble Matter ≤0.3 %
Assay (NH₂C(:NH)NH₂·HCl) 99.5 - 101.0 %
Ultraviolet Absorbance - 275 nm, For Information Only
Ultraviolet Absorbance - 260 nm, For Information Only
Loss on Drying ≤0.5 %
Trace Impurities - Iron (Fe) ≤2.0 ppm
MANUFACTURED EXCLUSIVELY FOR AVANTOR PERFORMANCE MATERIALS, LLC., BY
BIOSPECTRA, INC., STROUDSBURG, PA

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR