Calcium sulfate, Drierite™ 3 mm (8 mesh) for drying, without indicator, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

L056-02 L056-07
CAJTL056-2EA 1946.21 CAD
CAJTL056-2 CAJTL056-7 JTL056-2 JTL056-7
Calcium sulfate, Drierite™ 3 mm (8 mesh) for drying, without indicator, J.T.Baker®
Calcium sulphate
Formula: CaO₄S
Melting Pt: 1350 °C
Density: 2.964 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3
Merck Index: 13,01711

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR