Sulfuric acid ≥95% (by acidimetry) NF, FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

BAKR9675-03 BAKR9675-07 BAKR9675-08 9675-02 BAKR9675-25 BAKR9675-R
CAJT9675-3CS 2498.17 CAD
CAJT9675-3 CAJT9675-7 CAJT9675-8 JT9675-2 CAJT9675-25 CAJT9675-R
Sulfuric acid ≥95% (by acidimetry) NF, FCC, J.T.Baker®
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Meets N.F. Requirements
Meets F.C.C. Requirements
GMP Manufactured Product
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
NF - Assay (H₂SO₄) (by acidimetry) 95.0 - 98.0 %
NF - Arsenic (As) ≤1 ppm
NF - Chloride (Cl) ≤0.005 %
NF - Identification Passes Test
NF - Reducing Substances Passes Test
NF - Residue on Ignition ≤0.005 %
FCC - Assay (H₂SO₄) (by acidimetry) 95.0 - 98.0 %
FCC - Arsenic (As) ≤1 ppm
FCC - Chloride (Cl) ≤0.005 %
FCC - Iron (Fe) ≤0.02 %
FCC - Identification Passes Test
FCC - Lead (Pb) ≤5 mg/kg
FCC - Nitrate (NO₃) ≤10 mg/kg
FCC - Reducing Substances Passes Test
FCC - Selenium (Se) ≤0.002 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR