Sulfuric acid ≥95% (by acidimetry) NF, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

BAKR9671-03 9671-02 BAKR9671-26
CAJT9671-3CS 2432.52 CAD
CAJT9671-3 JT9671-2 CAJT9671-26 CAJT9671-2
Sulfuric acid ≥95% (by acidimetry) NF, Multi-Compendial, J.T.Baker®
Sulfuric acid
Formula: H₂SO₄
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Identification Passes Test
NF - Residue on Ignition ≤ 0.005 %
NF - Chloride (Cl) ≤ 0.005 %
NF - Arsenic (As) ≤ 1 ppm
NF - Reducing Substances Passes Test
NF - Assay (as H₂SO₄) (by acidimetry) 95.0 - 98.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Chloride (Cl) ≤ 50 ppm
EP/BP - Nitrate (NO₃) Passes Test
EP/BP - Iron (Fe) ≤ 25 ppm
EP/BP - Assay (H₂SO₄) 95.0 - 100.5 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR