Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Ethyl nitrileMethyl cyanideEthanoic acid nitrileEthanenitrileCyanomethane

Management of Change (MOC) category = N

9012-03 BAKR9012-99 BAKR9012-03 9012-QM 9012-ZX 9012-99 BAKR9012-QM 9012-98 BAKR9012-ZX
CAJT9012-QMCS 78.02 CAD
JT9012-QM CAJT9012-QM JT9012-ZX JT9012-3 CAJT9012-ZX JT9012-99 CAJT9012-3 CAJT9012-99 CAJT9012-98
Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®
Acetonitrile
Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
MDL Number: MFCD00001878
CAS Number: 75-05-8
UN: 1648
ADR: 3,II
Merck Index: 13,00071
https://ca.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (GC) ≥99.9 %
Color (APHA) ≤10
Water (H₂O) ≤0.02 %
Gradient Elution Test (a.u.) - 254 nm ≤0.005
Residue after Evaporation ≤3 ppm
Titrable Acid (µeq/g) ≤8
Titrable Base (µeq/g) ≤0.6
Ultraviolet Absorbance (1.00 cm) - 200 nm ≤0.1
Ultraviolet Absorbance (1.00 cm) - 210 nm ≤0.05
Ultraviolet Absorbance (1.00 cm) - 220 nm ≤0.03
Ultraviolet Absorbance (1.00 cm) - 254 nm ≤0.01
Ultraviolet Absorbance (1.00 cm) - 280 nm ≤0.01
Ultraviolet Absorbance (1.00 cm) - 350 nm ≤0.01
Ultraviolet Absorbance (1.00 cm) - 400 nm ≤0.01
Ultraviolet Absorbance (1.00 cm) - UV Cut-off, nm ≤190
Filtered through a 0.2 micron filter.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR