EDTA disodium salt dihydrate 99,0-101,0 Dried, crystals USP, multi-compendial

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

M.O.C. Category R

BAKR8995-20 BAKR8995-07 BAKR8995-01
CAJT8995-20EA 15426.58 CAD
CAJT8995-20 CAJT8995-7 CAJT8995-1
EDTA disodium salt dihydrate 99,0-101,0 Dried, crystals USP, multi-compendial
EDTA disodium salt dihydrate
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Melting Pt: 255
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets J.P. Chemical Specifications
Meets U.S.P Requirements
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
USP - pH (1 in 20) 4.0 - 6.0
USP - Loss on Drying 8.7 - 11.4 %
USP - Calcium (Ca) Passes Test
USP - Nitrilotriacetic Acid ≤0.1 %
USP - Assay (C₁₀H₁₄N₂Na₂O₈) (dried basis) 99.0 - 101.0 %
Endotoxin Concentration (EU/g) ≤5
EP/BP - Assay 98.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification D Passes Test
EP/BP - Impurity A ≤0.1 %
EP/BP - Appearance of Solution Passes Test
EP/BP - pH (1 in 20) 4.0 - 5.5
EP/BP - Iron (Fe) ≤80 ppm
JP - Assay 99.0 - 101.0 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Identification C Passes Test
JP - pH of 1% Solution at 25°C 4.3 - 4.7
JP - Clarity and Color of Solution Passes Test
JP - Cyanide (CN) Passes Test
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Arsenic (As) ≤2 ppm
JP - Residue after Ignition 37.0 - 39.0 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR