Sodium thiosulfate, anhydrous ≥98.0%, granular, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

3953-01 BAKR3953-05 BAKR3953-09
JT3953-1CS 669.35 CAD
JT3953-1 CAJT3953-5 CAJT3953-9
Sodium thiosulfate, anhydrous ≥98.0%, granular, BAKER ANALYZED®, J.T.Baker®
Sodium thiosulphate
Formula: Na₂S₂O₃
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 48 °C
Density: 1.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (Na₂S₂O₃) ≥98.0 %
Insoluble Matter ≤0.01 %
Calcium (Ca) ≤0.01 %
Heavy Metals (as Pb) ≤0.001 %
Sulfide (S) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR