Disodium hydrogen phosphate dodecahydrate 98.0-102.0%, crystals, BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
sec-Sodium phosphate, Sodium hydrogen phosphate dodecahydrate, Disodium phosphate, Dibasic sodium phosphate dodecahydrate, Sodium phosphate dibasic dodecahydrate, di-Sodium hydrogen phosphate dodecahydrate, Sodium monohydrogen phosphate dodecahydrate, di-Sodium hydrogen orthophosphate dodecahydrate
Management of Change (MOC) category = N
BAKR3822-07
BAKR3822-01
BAKR3822-05
CAJT3822-7EA
2320.42
CAD
CAJT3822-7
CAJT3822-1
CAJT3822-5
Disodium hydrogen phosphate dodecahydrate 98.0-102.0%, crystals, BAKER ANALYZED®, J.T.Baker®
di-Sodium hydrogen phosphate dodecahydrate
Formula:
Na₂HPO₄·12H₂O MW: 358.14 g/mol Melting Pt: 35 Density: 1.52 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149181 CAS Number: 10039-32-4 EINECS: 231-448-7 Merck Index: 13,08733 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay | 98.0 - 102.0 % |
Insoluble Matter | ≤0.005 % |
pH of 5% Solution at 25°C | 8.7 - 9.3 |
Chloride (Cl) | ≤0.001 % |
Sulfate (SO₄) | ≤0.002 % |
Heavy Metals (as Pb) | ≤0.001 % |
Trace Impurities - Iron (Fe) | ≤0.001 % |
Nitrogen Compounds (as N) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...