Sodium benzoate 99 Anhydrous basis NF, multi-compendial

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Benzoic acid sodium salt

M.O.C. Category R

BAKR3501-05 BAKR3501-20
CAJT3501-5EA 856.85 CAD
CAJT3501-5 CAJT3501-20
Sodium benzoate 99 Anhydrous basis NF, multi-compendial
Sodium benzoate
Formula: C₆H₅COONa
MW: 144.11 g/mol
Melting Pt: 410…430 °C
Density: 1.44 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 13,08654

Order Now

Specification Test Results

GMP Manufactured Product
Meets E.P. Chemical Specifications
Meets N.F. Requirements
Meets B.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Identification C Passes Test
NF - Alkalinity Passes Test
NF - Water (H₂O) ≤ 1.5 %
NF - Assay (C₆H₅COONa) (anhydrous basis) 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Halogenated Compounds (as Cl) - Ionized Chlorine ≤ 200 ppm
EP/BP - Halogenated Compounds (as Cl) - Total Chlorine ≤ 300 ppm
EP/BP - Loss on Drying ≤ 2.0 %
EP/BP - Assay(C₆H₅COONa)(dried basis) 99.0 - 100.5 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR