Silica gel, BAKER ANALYZED® 60-200 mesh for column chromatography, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

BAKR3414-01 BAKR3414-05
CAJT3414-1CS 5093.68 CAD
CAJT3414-1 CAJT3414-5
Silica gel, BAKER ANALYZED® 60-200 mesh for column chromatography, J.T.Baker®
Silica gel
Formula: SiO₂
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
REACH: 01-2119379499-16

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Toluene Absorption Passes Test
Loss on Ignition ≤10.0 %
Alcohol-Ether Soluble Substances ≤0.10 %
Bulk Density (g/cc), For Information Only
Mesh - On U.S. No. 60 Sieve ≤15 %
Mesh - Thru U.S. No. 200 Sieve ≤15 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR