L(+)-Potassium sodium tartrate tetrahydrate 99, crystals, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Potassium sodium tartrate tetrahydrate, Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate, Potassium sodium L(+)-tartrate tetrahydrate, Sodium potassium tartrate, (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrate, (R,R)-(+)-Potassium sodium tartrate tetrahydrate, Rochelle salt, L(+)-Tartaric acid potassium sodium salt tetrahydrate, (+)-Potassium sodium L-tartrate tetrahydrate

BAKR3262-07 BAKR3262-01 CAJT3262-19 BAKR3262-05
CAJT3262-7EA 3371.47 CAD
CAJT3262-7 CAJT3262-1 CAJT3262-19 CAJT3262-5
L(+)-Potassium sodium tartrate tetrahydrate 99, crystals, BAKER ANALYZED® ACS
L(+)-Potassium sodium tartrate tetrahydrate
Formula: KOOC(CHOH)₂COONa·4H₂O
MW: 282.22 g/mol
Melting Pt: 70…80 °C
Density: 1.79 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS Number: 6381-59-5
EINECS: 206-156-8
Merck Index: 13,07755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d585522f4e3b14a7b011dc8a858f5eeca413864&vid=542d22225b124dda886e5007091b35db67214bb7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (KOCO(CHOH)₂COONa·4H₂O) 99.0 - 102.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 6.0 - 8.5
Chloride (Cl) ≤0.001 %
Phosphate (PO₄) ≤0.002 %
Sulfate (SO₄) ≤0.005 %
Ammonium (NH₄) ≤0.002 %
Calcium (Ca) ≤0.005 %
Trace Impurities - Iron (Fe) ≤0.001 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR