Potassium nitrate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = NI
BAKR3190-07
BAKR3190-80
CAJT3190-7EA
1757.92
CAD
CAJT3190-7
CAJT3190-1
CAJT3190-5
JT3190-1
CAJT3190-80
Potassium nitrate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium nitrate
Formula:
KNO₃ Melting Pt: 334 °C Density: 2.109 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011409 CAS Number: 7757-79-1 EINECS: 231-818-8 UN: 1486 ADR: 5.1,III REACH: 01-2119488224-35 Merck Index: 13,07733 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (KNO₃) | ≥99.0 % |
Insoluble Matter | ≤0.005 % |
pH of 5% Solution at 25°C | 4.5 - 8.5 |
Chloride (Cl) | ≤0.002 % |
ACS- Nitrite (NO₂) | ≤0.001 % |
ACS - Sulfate (SO₄) | ≤0.003 % |
Calcium (Ca) | ≤0.005 % |
ACS - Magnesium (Mg) | ≤0.002 % |
Sodium (Na) | ≤0.005 % |
Iodate (IO₃) | ≤5 ppm |
Phosphate (PO₄) | ≤3 ppm |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤2.0 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...