Potassium carbonate, anhydrous ≥99%, granules, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Potash

Management of Change (MOC) category = NI

BAKR3012-07 BAKR3012-09 BAKR3012-R BAKR3012-01 BAKR3012-05
CAJT3012-7EA 3597.36 CAD
CAJT3012-7 CAJT3012-9 CAJT3012-R CAJT3012-1 CAJT3012-5
Potassium carbonate, anhydrous ≥99%, granules, BAKER ANALYZED® ACS, J.T.Baker®
Potassium carbonate
Formula: K₂CO₃
MW: 138.21 g/mol
Melting Pt: 891
Density: 2.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (K₂CO₃) (by acid-base titration) ≥99.0 %
Insoluble Matter ≤0.01 %
Calcium (Ca) ≤0.002 %
Chloride (Cl) ≤0.003 %
ACS - Magnesium (Mg) ≤0.002 %
Phosphate (PO₄) ≤0.001 %
Silica (SiO₂) ≤0.005 %
Sulfur Compounds (as SO₄) ≤0.004 %
Sodium (Na) ≤0.02 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR