Potassium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = N
BAKR2997-01
BAKR2997-07
CAJT2997-1CS
3622.61
CAD
CAJT2997-1
CAJT2997-7
Potassium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium bromide
Formula:
KBr MW: 119 g/mol Boiling Pt: 1435 Melting Pt: 730 Density: 2.75 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011358 CAS Number: 7758-02-3 EINECS: 231-830-3 Merck Index: 13,07701 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (KBr) (by Ag titrn) | ≥99.0 % |
Insoluble Matter | ≤0.005 % |
pH of 5% Solution at 25°C | 5.0 - 8.8 |
Bromate (BrO₃) | ≤0.001 % |
Iodate (IO₃) | ≤0.001 % |
Chloride (Cl) | ≤0.2 % |
Iodide (I) | ≤0.001 % |
Nitrogen Compounds (as N) | ≤0.005 % |
Sulfate (SO₄) | ≤0.005 % |
Barium (Ba) | ≤0.002 % |
Calcium (Ca) | ≤0.002 % |
Magnesium (Mg) | ≤0.001 % |
Sodium (Na) | ≤0.02 % |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤5 ppm |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...