Manganese(II) chloride tetrahydrate ≥98% (by EDTA titration), crystals, BAKER ANALYZED® ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Synonyms:
Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate
Management of Change (MOC) category = N
BAKR2540-05
BAKR2540-07
BAKR2540-R
BAKR2540-01
CAJT2540-5EA
1048.69
CAD
CAJT2540-5
CAJT2540-7
CAJT2540-R
CAJT2540-1
Manganese(II) chloride tetrahydrate ≥98% (by EDTA titration), crystals, BAKER ANALYZED® ACS
Manganese (II) chloride tetrahydrate
Formula:
MnCl₂·4H₂O MW: 197.9 g/mol Boiling Pt: 1190 Melting Pt: 58…59 °C Density: 2.01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149792 CAS Number: 13446-34-9 EINECS: 231-869-6 UN: 3077 ADR: 9 III Merck Index: 13,05751 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (MnCl₂·4H₂O) (by EDTA titrn) | 98.0 - 101.0 % |
ACS - Insoluble Matter | ≤0.005 % |
ACS - pH of 5% Solution at 25°C | 3.5 - 6.0 |
ACS - Sulfate (SO₄) | ≤0.005 % |
ACS - Calcium (Ca) | ≤0.005 % |
ACS - Magnesium (Mg) | ≤0.005 % |
ACS - Potassium (K) | ≤0.01 % |
ACS - Sodium (Na) | ≤0.05 % |
ACS - Zinc (Zn) | ≤0.005 % |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...