Magnesium chloride hexahydrate 99-102%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Magnesium dichloride

Management of Change (MOC) category = N

BAKR2444-07 BAKR2444-01 2444-01 BAKR2444-R
CAJT2444-7EA 2924.49 CAD
CAJT2444-7 CAJT2444-1 CAJT2444-5 JT2444-1 CAJT2444-R
Magnesium chloride hexahydrate 99-102%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Magnesium chloride hexahydrate
Formula: MgCl₂·6H₂O
MW: 203.3 g/mol
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (MgCl₂·6H₂O) 99.0 - 102.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 4.5 - 7.0
Nitrate (NO₃) ≤0.001 %
Sulfate (SO₄) ≤0.002 %
Ammonium (NH₄) ≤0.002 %
Barium (Ba) ≤0.005 %
Calcium (Ca) ≤0.01 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Strontium (Sr) ≤0.005 %
Phosphate (PO₄) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Trace Impurities - Manganese (Mn) ≤5.0 ppm
The net weight of the product in each container is within +/- 2% of the
netweight stated on the label.
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR