Copper(II) sulfate pentahydrate USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Cupric sulfate pentahydrateCopper sulfate pentahydrateCopper sulphate pentahydrateCupric sulphate pentahydrate

Management of Change (MOC) category = RL

1846-07 BAKR1846-07
CAJT1846-7EA 5331.39 CAD
JT1846-7 CAJT1846-7
Copper(II) sulfate pentahydrate USP, Multi-Compendial, J.T.Baker®
Copper (II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Density: 2.284 g/cm³ (20 °C)
MDL Number: MFCD00149681
CAS Number: 7758-99-8
UN: 3077
ADR: 9,III
Merck Index: 13,02682

Order Now

Specification Test Results

Meets B.P. Chemical Specifications
Meets U.S.P Requirements
GMP Manufactured Product
Meets E.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Calcium (Ca) ≤0.005 %
USP - Identification Passes Test
USP - Iron (Fe) ≤0.003 %
USP - Loss on Drying 33.0 - 36.5 %
USP - Nickel (Ni) ≤0.005 %
USP - Potassium (K) ≤0.01 %
USP - Sodium (Na) ≤0.02 %
USP - Assay (CuSO₄) (dried basis) 98.5 - 100.5 %
Endotoxin Concentration (EU/g) ≤10
EP/BP - Assay 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Chloride (Cl) ≤100 ppm
EP/BP - Iron (Fe) ≤100 ppm
EP/BP - Lead (Pb) ≤50 ppm
EP/BP - Loss on Drying 35.0 - 36.5 %
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR