Sodium hydroxide solution 50%, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

BAKR0897-07 BAKR0897-09 BAKR0897-79
CAJT0897-07EA 5031.53 CAD
CAJT0897-07 CAJT0897-09 CAJT0897-79
Sodium hydroxide solution 50%, Biotech Reagent, J.T.Baker®
Sodium hydroxide

Made from Water for Injection (WFI) U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications

Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

GMP Manufactured Product
Store above 16°C (60°F) to prevent freezing
CAUTION: For Manufacturing, processing or repackaging
Assay (NaOH) 50 - 52 %
Chloride (Cl) ≤30 ppm
Heavy Metals (as Pb) ≤1 ppm
Iron (Fe) ≤2 ppm
Made from Water for Injection U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 5 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR