Ammonium sulfate ≥99% NF endotoxin tested, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

Management of Change (MOC) category = R

BAKR0798-F7 BAKR0798-01 BAKR0798-05 BAKR0798-65 BAKR0798-08 BAKR0798-F9
CAJT0798-F7EA 7247.27 CAD
CAJT0798-F7 CAJT0798-01 CAJT0798-05 CAJT0798-65 CAJT0798-8 CAJT0798-F9
Ammonium sulfate ≥99% NF endotoxin tested, J.T.Baker®
Ammonium sulphate

Preserve in well closed container.

Formula: (NH₄)₂SO₄
Melting Pt: 336…339 °C
Density: 1.77 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay ((NH₄)₂SO₄) 99.0 - 100.5 %
NF - Insoluble Matter ≤0.005 %
NF - Residue on Ignition ≤0.005 %
NF - pH of 5% Solution 5.0 - 6.0
NF - Nitrate (NO₃) ≤0.001 %
NF - Chloride (Cl) ≤5 ppm
NF - Phosphate (PO₄) ≤5 ppm
NF - Iron (Fe) ≤5 ppm
NF - Identification Passes Test
NF - Microbial Limit - Total Aerobic Microbial Count (cfu/g) ≤1000
NF - Microbial Limit - Total Combined Molds and Yeasts (cfu/g) ≤10
Endotoxin Concentration (EU/g) ≤2.5
No Class 1, 2 or 3 Residual Solvents are likely to be present by exclusion of the production/purification process.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.
Preserve in well closed container.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR