Sodium hydroxide 1.0 N in aqueous solution, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

BAKR0389-07 BAKR0389-09
CAJT0389-7EA 3010.56 CAD
CAJT0389-7 CAJT0389-9
Sodium hydroxide 1.0 N in aqueous solution, Biotech Reagent, J.T.Baker®
Sodium hydroxide

Made from water for injection USP and sodium hydroxide pellets, NF (multi-compendial) which meets BP, Ph. Eur. and JP chemical specifications.

Formula: NaOH
Density: 1.04 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 3266
ADR: 8,III

Order Now

Specification Test Results

GMP Manufactured Product
Protect from air to avoid absorption of carbon dioxide.
CAUTION: For Manufacturing, processing or repackaging
Appearance (Clear, colorless liquid) Passes Test
Normality 0.9 - 1.1
Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Endotoxin Concentration (EU/mL ≤2
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Made from Water for Injection U.S.P. and Sodium Hydroxide pellets, N.F.(Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
Filtered through a 0.2 micron filter.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR