L(+)-Tartaric acid ≥99.0% (by acidimetry), crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid

Management of Change (MOC) category = N

BAKR0386-07 BAKR0386-01 BAKR0386-05
CAJT0386-7EA 3436.23 CAD
CAJT0386-7 CAJT0386-1 CAJT0386-5
L(+)-Tartaric acid ≥99.0% (by acidimetry), crystals, BAKER ANALYZED® ACS, J.T.Baker®
L(+)-Tartaric acid

  • Lot analysis on label

Formula: C₄H₆O₆
MW: 150.09 g/mol
Boiling Pt: 399 °C (1013 hPa)
Melting Pt: 168…170 °C
Density: 1.759 g/cm³ (20 °C)
Flash Pt: 150 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00064207
CAS Number: 87-69-4
EINECS: 201-766-0
REACH: 01-2119537204-47
Merck Index: 13,09158
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (HOCO(CHOH)₂COOH) (by acidimetry) ≥99.0 %
Insoluble Matter ≤0.005 %
Residue after Ignition ≤0.020 %
Chloride (Cl) ≤0.001 %
Oxalate (C₂O₄) Passes Test
Phosphate (PO₄) ≤0.001 %
Sulfur Compounds (as S) ≤0.002 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR