Pyrogallol, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: VWR International
Warning

Synonyms: Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene

Management of Change (MOC) category = N

BAKR0289-01 BAKR0289-04
CAJT0289-1EA 1346.81 CAD
CAJT0289-1 CAJT0289-4
Pyrogallol, BAKER ANALYZED® ACS, J.T.Baker®
Pyrogallol
Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 309
Melting Pt: 130…135 °C
Density: 1.453 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002192
CAS Number: 87-66-1
EINECS: 201-762-9
UN: 2811
ADR: 6.1 III
Merck Index: 13,08090
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09c5271be2848ebfa6e3614bff56b329a59cb6fe&vid=db25c87d92a56d9ff5d36f04d6bbf59acf236659&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Melting Point 131.0 - 135.0 °C
Residue after Ignition ≤0.005 %
Chloride (Cl) ≤0.001 %
Sulfate (SO₄) ≤0.005 %
Trace Impurities - Iron (Fe) ≤0.001 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR