Citric acid monohydrate 99.0-102.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

Management of Change (MOC) category = N

BAKR0117-01 BAKR0117-05
CAJT0117-1CS 1387.76 CAD
CAJT0117-1 CAJT0117-5
Citric acid monohydrate 99.0-102.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
Citric acid monohydrate
Formula: C₆H₈O₇.H₂O
Melting Pt: 135…152 °C
Density: 1.552 g/cm³ (20 °C)
Flash Pt: 173.9 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00149972
CAS Number: 5949-29-1
EINECS: 201-069-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (HOC(COOH)(CH₂COOH)₂·H₂O) (by acidimetry) 99.0 - 102.0 %
Insoluble Matter ≤0.005 %
Residue on Ignition ≤0.02 %
Chloride (Cl) ≤0.001 %
Oxalate (C₂O₄) Passes Test
Phosphate (PO₄) ≤0.001 %
Sulfur Compounds (as SO₄) ≤0.002 %
Substances Carbonizable by Hot H₂SO₄ Passes Test
Trace Impurities - Iron (Fe) ≤2.0 ppm
Trace Impurities - Lead (Pb) ≤2.000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR